siRNA Details
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012856.1 | NP_036988.1 | 2 | 1821 | ||
GenBank mRNA | M68880.1 | - | 2 | 1821 | ||
X62840.1 | - | 2 | 946 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012856.1 | NP_036988.1 | 2 | 1821 | ||
GenBank mRNA | M68880.1 | - | 2 | 1821 | ||
X62840.1 | - | 2 | 946 |