siRNA Details
| Gene Symbol: Nfyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031553.2 | NP_113741.1 | 3 | 269 | ||
| XM_006241196.2 | XP_006241258.1 | 3 | 287 | |||
| GenBank mRNA | BC089791.1 | - | 3 | 269 | 7380395 | |
| M55045.1 | - | 2 | 138 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfyb| Gene Symbol: Nfyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031553.2 | NP_113741.1 | 3 | 269 | ||
| XM_006241196.2 | XP_006241258.1 | 3 | 287 | |||
| GenBank mRNA | BC089791.1 | - | 3 | 269 | 7380395 | |
| M55045.1 | - | 2 | 138 | |||