siRNA Details
| Gene Symbol: Nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012866.2 | NP_036998.1 | 2, 3 | 310 | ||
| XM_006238785.2 | XP_006238847.1 | 2, 3 | 283 | |||
| GenBank mRNA | BC093619.1 | - | 2, 3 | 310 | 7314492 | |
| U17607.1 | - | 2, 3 | 198 | |||

Nfyc
Genomic Map