siRNA Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012868.1 | NP_037000.1 | 8 | 1590 | ||
XM_006232045.2 | XP_006232107.1 | 8 | 2177 | |||
GenBank mRNA | L27339.1 | - | 8 | 1590 | ||
X75605.1 | - | 8 | 1611 | |||
X78595.1 | - | 1436 |
