siRNA Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012868.1 | NP_037000.1 | 4 | 1102 | ||
XM_006232045.2 | XP_006232107.1 | 4 | 1686 | |||
GenBank mRNA | L27339.1 | - | 4 | 1102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012868.1 | NP_037000.1 | 4 | 1102 | ||
XM_006232045.2 | XP_006232107.1 | 4 | 1686 | |||
GenBank mRNA | L27339.1 | - | 4 | 1102 |