siRNA Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012881.2 | NP_037013.2 | 7 | 742 | ||
| XM_008769996.1 | XP_008768218.1 | 7 | 815 | |||
| GenBank mRNA | AB001382.1 | - | 6 | 636 | ||
| BC078874.1 | - | 7 | 722 | 7110677 | ||
| FQ213473.1 | - | 7 | 731 | |||
| FQ222192.1 | - | 7 | 758 | |||
| M14656.1 | - | 7 | 715 | |||
| M99252.1 | - | 7 | 751 | |||

Spp1
Genomic Map