siRNA Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012881.2 | NP_037013.2 | 7 | 935 | ||
| XM_008769996.1 | XP_008768218.1 | 7 | 1008 | |||
| GenBank mRNA | AB001382.1 | - | 6 | 829 | ||
| BC078874.1 | - | 7 | 915 | 7110677 | ||
| BU759036.1 | - | 1 | 565 | |||
| FQ213473.1 | - | 7 | 924 | |||
| FQ222192.1 | - | 7 | 951 | |||
| M14656.1 | - | 7 | 908 | |||
| M99252.1 | - | 7 | 944 | |||

Spp1
Genomic Map