siRNA Details
| Gene Symbol: Vcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012889.1 | NP_037021.1 | 1698 | |||
| GenBank mRNA | BC089812.1 | - | 1716 | 7364929 | ||
| M84488.1 | - | 1698 | ||||
| X63722.1 | - | 1647 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcam1| Gene Symbol: Vcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012889.1 | NP_037021.1 | 1698 | |||
| GenBank mRNA | BC089812.1 | - | 1716 | 7364929 | ||
| M84488.1 | - | 1698 | ||||
| X63722.1 | - | 1647 | ||||