siRNA Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083122.1 | NP_001076591.1 | 6, 7 | 908 | ||
| NM_012890.2 | NP_037022.1 | 5, 6 | 847 | |||
| GenBank mRNA | BC061997.1 | - | 5, 6 | 850 | ||
| BC168237.1 | - | 6, 7 | 901 | 7938890 | ||
| U50927.1 | - | 5, 6 | 847 | |||

Slc30a2
Genomic Map