siRNA Details
Gene Symbol: Aldh3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031972.1 | NP_114178.1 | 2 | 185 | ||
GenBank mRNA | BC070924.1 | - | 2 | 218 | 7096634 | |
J03637.1 | - | 2 | 185 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aldh3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031972.1 | NP_114178.1 | 2 | 185 | ||
GenBank mRNA | BC070924.1 | - | 2 | 218 | 7096634 | |
J03637.1 | - | 2 | 185 |