siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012910.2 | NP_037042.1 | 5, 6 | 383 | ||
| XM_006229734.2 | XP_006229796.1 | 5, 6 | 1009 | |||
| XM_006229735.2 | XP_006229797.1 | 5, 6 | 1008 | |||
| XM_006229736.2 | XP_006229798.1 | 5, 6 | 978 | |||
| XM_006229737.2 | XP_006229799.1 | 5, 6 | 977 | |||
| XM_006229738.2 | XP_006229800.1 | 5, 6 | 1010 | |||
| XM_006229739.2 | XP_006229801.1 | 5, 6 | 1125 | |||
| XM_006229740.2 | XP_006229802.1 | 5, 6 | 511 | |||
| XM_006229741.2 | XP_006229803.1 | 5, 6 | 524 | |||
| GenBank mRNA | M91589.1 | - | 5, 6 | 386 | ||

Arrb1
Genomic Map