siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012910.2 | NP_037042.1 | 8, 9 | 642 | ||
| XM_006229734.2 | XP_006229796.1 | 8, 9 | 1268 | |||
| XM_006229735.2 | XP_006229797.1 | 8, 9 | 1267 | |||
| XM_006229736.2 | XP_006229798.1 | 8, 9 | 1237 | |||
| XM_006229737.2 | XP_006229799.1 | 8, 9 | 1236 | |||
| XM_006229738.2 | XP_006229800.1 | 8, 9 | 1269 | |||
| XM_006229739.2 | XP_006229801.1 | 8, 9 | 1384 | |||
| XM_006229740.2 | XP_006229802.1 | 8, 9 | 770 | |||
| XM_006229741.2 | XP_006229803.1 | 8, 9 | 783 | |||
| GenBank mRNA | M91589.1 | - | 8, 9 | 645 | ||

Arrb1
Genomic Map