siRNA Details
| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031555.1 | NP_113743.1 | 8 | 1882 | ||
| XM_006235117.2 | XP_006235179.1 | 8 | 1543 | |||
| GenBank mRNA | AB003104.1 | - | 8 | 1883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bfsp1| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031555.1 | NP_113743.1 | 8 | 1882 | ||
| XM_006235117.2 | XP_006235179.1 | 8 | 1543 | |||
| GenBank mRNA | AB003104.1 | - | 8 | 1883 | ||