siRNA Details
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012920.1 | NP_037052.1 | 9 | 657 | ||
GenBank mRNA | J02942.1 | - | 9 | 657 | ||
M16960.1 | - | 3 | 223 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012920.1 | NP_037052.1 | 9 | 657 | ||
GenBank mRNA | J02942.1 | - | 9 | 657 | ||
M16960.1 | - | 3 | 223 |