siRNA Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012927.1 | NP_037059.1 | 3 | 476 | ||
| GenBank mRNA | D25290.1 | - | 3 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh6| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012927.1 | NP_037059.1 | 3 | 476 | ||
| GenBank mRNA | D25290.1 | - | 3 | 476 | ||