siRNA Details
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012932.2 | NP_037064.1 | 8, 9 | 1021 | ||
GenBank mRNA | U52102.1 | - | 8, 9 | 989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012932.2 | NP_037064.1 | 8, 9 | 1021 | ||
GenBank mRNA | U52102.1 | - | 8, 9 | 989 |