siRNA Details
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012933.1 | NP_037065.1 | 10 | 1179 | ||
XM_008759973.1 | XP_008758195.1 | 7 | 790 | |||
GenBank mRNA | U52103.1 | - | 9 | 1042 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012933.1 | NP_037065.1 | 10 | 1179 | ||
XM_008759973.1 | XP_008758195.1 | 7 | 790 | |||
GenBank mRNA | U52103.1 | - | 9 | 1042 |