siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012934.1 | NP_037066.1 | 1, 2 | 121 | ||
| GenBank mRNA | AF389425.1 | - | 1, 2 | 121 | ||
| U52104.1 | - | 1, 2 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpysl3| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012934.1 | NP_037066.1 | 1, 2 | 121 | ||
| GenBank mRNA | AF389425.1 | - | 1, 2 | 121 | ||
| U52104.1 | - | 1, 2 | 337 | |||