siRNA Details
| Gene Symbol: Ctse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012938.1 | NP_037070.1 | 7, 8 | 983 | ||
| XM_006249748.2 | XP_006249810.1 | 7, 8 | 1188 | |||
| XM_006249749.2 | XP_006249811.1 | 8, 9 | 1246 | |||
| GenBank mRNA | BC062002.1 | - | 7, 8 | 993 | 5599861 | |
| D38104.1 | - | 7, 8 | 983 | |||
| D45187.1 | - | 7, 8 | 1125 | |||

Ctse
Genomic Map