siRNA Details
| Gene Symbol: Cyp7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012942.2 | NP_037074.1 | 6 | 1380 | ||
| GenBank mRNA | J05430.1 | - | 6 | 1375 | ||
| J05460.1 | - | 6 | 1381 | |||
| J05509.1 | - | 6 | 1392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp7a1| Gene Symbol: Cyp7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012942.2 | NP_037074.1 | 6 | 1380 | ||
| GenBank mRNA | J05430.1 | - | 6 | 1375 | ||
| J05460.1 | - | 6 | 1381 | |||
| J05509.1 | - | 6 | 1392 | |||