siRNA Details
| Gene Symbol: Cyp7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019138.1 | NP_062011.1 | 3 | 792 | ||
| GenBank mRNA | U36992.1 | - | 1 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp7b1| Gene Symbol: Cyp7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019138.1 | NP_062011.1 | 3 | 792 | ||
| GenBank mRNA | U36992.1 | - | 1 | 395 | ||