siRNA Details
| Gene Symbol: Dio1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021653.3 | NP_067685.5 | 4 | 710 | ||
| GenBank mRNA | BC083557.1 | - | 4 | 705 | 7110283 | |
| X57999.1 | - | 4 | 709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dio1| Gene Symbol: Dio1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021653.3 | NP_067685.5 | 4 | 710 | ||
| GenBank mRNA | BC083557.1 | - | 4 | 705 | 7110283 | |
| X57999.1 | - | 4 | 709 | |||