siRNA Details
Gene Symbol: Dlx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012943.1 | NP_037075.1 | 2 | 632 | ||
GenBank mRNA | AB073716.1 | - | 2 | 354 | ||
D31734.1 | - | 2 | 629 | |||
L24443.1 | - | 2 | 632 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dlx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012943.1 | NP_037075.1 | 2 | 632 | ||
GenBank mRNA | AB073716.1 | - | 2 | 354 | ||
D31734.1 | - | 2 | 629 | |||
L24443.1 | - | 2 | 632 |