siRNA Details
| Gene Symbol: F2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012950.2 | NP_037082.2 | 2 | 398 | ||
| GenBank mRNA | BC085759.1 | - | 2 | 398 | 7106133 | |
| M81642.1 | - | 2 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2r| Gene Symbol: F2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012950.2 | NP_037082.2 | 2 | 398 | ||
| GenBank mRNA | BC085759.1 | - | 2 | 398 | 7106133 | |
| M81642.1 | - | 2 | 406 | |||