siRNA Details
| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012962.1 | NP_037094.1 | 10 | 1052 | ||
| XM_008762303.1 | XP_008760525.1 | 10 | 1466 | |||
| GenBank mRNA | BC078700.1 | - | 9 | 1017 | 7110971 | |
| L38615.1 | - | 10 | 1052 | |||

Gss
Genomic Map