siRNA Details
| Gene Symbol: Il1rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167840.1 | NP_001161312.1 | 3 | 344 | ||
| NM_012968.1 | NP_037100.1 | 3 | 263 | |||
| XM_006248504.2 | XP_006248566.1 | 3 | 332 | |||
| GenBank mRNA | BC097339.1 | - | 3 | 279 | 7442849 | |
| GU123169.1 | - | 3 | 344 | |||
| U48592.1 | - | 3 | 263 | |||

Il1rap
Genomic Map