siRNA Details
| Gene Symbol: Irs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012969.1 | NP_037101.1 | 1 | 2586 | ||
| GenBank mRNA | X58375.1 | - | 1 | 2586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irs1| Gene Symbol: Irs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012969.1 | NP_037101.1 | 1 | 2586 | ||
| GenBank mRNA | X58375.1 | - | 1 | 2586 | ||