siRNA Details
Gene Symbol: Irs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012969.1 | NP_037101.1 | 1 | 2202 | ||
GenBank mRNA | X58375.1 | - | 1 | 2202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012969.1 | NP_037101.1 | 1 | 2202 | ||
GenBank mRNA | X58375.1 | - | 1 | 2202 |