siRNA Details
| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012974.1 | NP_037106.1 | 25 | 3724 | ||
| XM_006243709.2 | XP_006243771.1 | 25 | 3769 | |||
| GenBank mRNA | BC088400.1 | - | 25 | 3723 | 7107264 | |
| X16563.1 | - | 25 | 3724 | |||

Lamb2
Genomic Map