siRNA Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012977.1 | NP_037109.1 | 6, 7 | 617 | ||
| GenBank mRNA | BC061566.1 | - | 6, 7 | 578 | 6921383 | |
| U59462.1 | - | 6, 7 | 539 | |||
| U67958.1 | - | 6, 7 | 617 | |||
| U72741.1 | - | 7, 8 | 635 | |||

Lgals9
Genomic Map