siRNA Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012977.1 | NP_037109.1 | 8 | 713 | ||
| GenBank mRNA | BC061566.1 | - | 8 | 674 | 6921383 | |
| U59462.1 | - | 8 | 635 | |||
| U67958.1 | - | 8 | 713 | |||
| U72741.1 | - | 9 | 731 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals9| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012977.1 | NP_037109.1 | 8 | 713 | ||
| GenBank mRNA | BC061566.1 | - | 8 | 674 | 6921383 | |
| U59462.1 | - | 8 | 635 | |||
| U67958.1 | - | 8 | 713 | |||
| U72741.1 | - | 9 | 731 | |||