siRNA Details
| Gene Symbol: Mras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012981.2 | NP_037113.2 | 2 | 256 | ||
| GenBank mRNA | BC085752.1 | - | 2 | 256 | 7130510 | |
| D89863.1 | - | 664 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mras| Gene Symbol: Mras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012981.2 | NP_037113.2 | 2 | 256 | ||
| GenBank mRNA | BC085752.1 | - | 2 | 256 | 7130510 | |
| D89863.1 | - | 664 | ||||