siRNA Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012983.2 | NP_037115.2 | 5 | 751 | ||
| XM_008767949.1 | XP_008766171.1 | 5 | 2520 | |||
| GenBank mRNA | BC087027.1 | - | 5 | 751 | 7106587 | |
| X71997.1 | - | 5 | 801 | |||

Myo1d
Genomic Map