siRNA Details
| Gene Symbol: P2rx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142367.2 | NP_001135839.1 | 1 | 241 | ||
| NM_001285415.1 | NP_001272344.1 | 1 | 241 | |||
| NM_012997.3 | NP_037129.1 | 1 | 241 | |||
| GenBank mRNA | AF231010.1 | - | 1 | 32 | ||
| BC061742.1 | - | 1 | 233 | 5599053 | ||
| FJ424511.1 | - | 1 | 32 | |||
| X80477.1 | - | 1 | 241 | |||

P2rx1
Genomic Map