siRNA Details
| Gene Symbol: P4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012998.1 | NP_037130.1 | 7 | 947 | ||
| GenBank mRNA | BC061857.1 | - | 7 | 961 | 5623688 | |
| M21018.1 | - | 7 | 825 | |||
| M21476.1 | - | 5 | 524 | |||
| X02918.1 | - | 7 | 947 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P4hb| Gene Symbol: P4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012998.1 | NP_037130.1 | 7 | 947 | ||
| GenBank mRNA | BC061857.1 | - | 7 | 961 | 5623688 | |
| M21018.1 | - | 7 | 825 | |||
| M21476.1 | - | 5 | 524 | |||
| X02918.1 | - | 7 | 947 | |||