siRNA Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013000.2 | NP_037132.2 | 14 | 1407 | ||
| XM_006245594.2 | XP_006245656.1 | 14 | 1620 | |||
| XM_006245595.2 | XP_006245657.1 | 14 | 1620 | |||
| XM_006245596.2 | XP_006245658.1 | 14 | 1620 | |||
| XM_006245597.2 | XP_006245659.1 | 14 | 1620 | |||
| XM_008767360.1 | XP_008765582.1 | 14 | 1620 | |||
| GenBank mRNA | M25732.1 | - | 14 | 1407 | ||
| X59685.1 | - | 11 | 881 | |||
| X59687.1 | - | 13 | 1049 | |||
| X59688.1 | - | 13 | 1038 | |||

Pam
Genomic Map