siRNA Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013001.2 | NP_037133.1 | 12 | 1408 | ||
| XM_006234633.2 | XP_006234695.1 | 14 | 1833 | |||
| XM_006234634.2 | XP_006234696.1 | 14 | 3289 | |||
| XM_006234635.2 | XP_006234697.1 | 14 | 1748 | |||
| XM_006234636.2 | XP_006234698.1 | 14 | 2739 | |||
| XM_008762060.1 | XP_008760282.1 | 13 | 1932 | |||
| GenBank mRNA | AJ627631.1 | - | 10 | 1290 | ||
| AY540905.1 | - | 11 | 1305 | |||
| AY540906.1 | - | 11 | 1305 | |||
| BC128741.1 | - | 12 | 1408 | 8363963 | ||
| U69644.1 | - | 12 | 1628 | |||

Pax6
Genomic Map