siRNA Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013016.2 | NP_037148.2 | 3 | 345 | ||
| XM_006234951.2 | XP_006235013.1 | 2 | 643 | |||
| XM_006234952.2 | XP_006235014.1 | 3 | 1005 | |||
| XM_006234953.2 | XP_006235015.1 | 3 | 1187 | |||
| GenBank mRNA | AF055065.1 | - | 3 | 209 | ||
| BC099773.1 | - | 3 | 281 | 7932298 | ||
| D38468.1 | - | 2 | 400 | |||
| D85183.1 | - | 2 | 157 | |||
| U62328.1 | - | 3 | 345 | |||

Sirpa
Genomic Map