siRNA Details
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013023.2 | NP_037155.2 | 12, 13 | 1142 | ||
| XM_008767196.1 | XP_008765418.1 | 12, 13 | 1405 | |||
| GenBank mRNA | M60737.1 | - | 11, 12 | 1033 | ||
| X15353.1 | - | 11, 12 | 1033 | |||
| X51781.1 | - | 12, 13 | 1142 | |||

Sag
Genomic Map