siRNA Details
| Gene Symbol: St8sia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013029.1 | NP_037161.1 | 1, 2 | 239 | ||
| GenBank mRNA | U55938.1 | - | 1, 2 | 239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia3| Gene Symbol: St8sia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013029.1 | NP_037161.1 | 1, 2 | 239 | ||
| GenBank mRNA | U55938.1 | - | 1, 2 | 239 | ||