siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127689.1 | NP_001121161.1 | 5 | 852 | ||
| NM_013037.1 | NP_037169.1 | 5 | 779 | |||
| XM_006244739.2 | XP_006244801.1 | 5 | 1854 | |||
| XM_006244740.2 | XP_006244802.1 | 6 | 1062 | |||
| XM_006244742.2 | XP_006244804.1 | 6 | 2275 | |||
| XM_008766992.1 | XP_008765214.1 | 6 | 1099 | |||
| GenBank mRNA | EU929068.1 | - | 1 | 51 | ||
| U04317.1 | - | 5 | 852 | |||
| U04319.1 | - | 5 | 779 | |||

Il1rl1
Genomic Map