siRNA Details
| Gene Symbol: Sycp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013041.1 | NP_037173.1 | 2 | 165 | ||
| GenBank mRNA | X75785.1 | - | 2 | 165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sycp3| Gene Symbol: Sycp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013041.1 | NP_037173.1 | 2 | 165 | ||
| GenBank mRNA | X75785.1 | - | 2 | 165 | ||