siRNA Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019141.2 | NP_062014.2 | 6 | 502 | ||
| XM_006231128.2 | XP_006231190.1 | 7 | 1188 | |||
| XM_006231129.2 | XP_006231191.1 | 6, 7 | 1185 | |||
| XM_008760286.1 | XP_008758508.1 | 5, 6 | 1110 | |||
| GenBank mRNA | CB717588.1 | - | 5 | 227 | ||
| L14463.1 | - | 6 | 498 | |||

Tle4
Genomic Map