siRNA Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019141.2 | NP_062014.2 | 10, 11 | 1026 | ||
| XM_006231128.2 | XP_006231190.1 | 11, 12 | 1712 | |||
| XM_006231129.2 | XP_006231191.1 | 11, 12 | 1709 | |||
| XM_008760286.1 | XP_008758508.1 | 10, 11 | 1634 | |||
| GenBank mRNA | L14463.1 | - | 10, 11 | 1019 | ||

Tle4
Genomic Map