siRNA Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019141.2 | NP_062014.2 | 17, 18 | 2240 | ||
| XM_006231128.2 | XP_006231190.1 | 18, 19 | 2926 | |||
| XM_006231129.2 | XP_006231191.1 | 18, 19 | 2923 | |||
| XM_008760286.1 | XP_008758508.1 | 17, 18 | 2848 | |||
| GenBank mRNA | CB766637.1 | - | 2, 3 | 316 | ||
| L14463.1 | - | 17, 18 | 2233 | |||

Tle4
Genomic Map