siRNA Details
Gene Symbol: Trhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013047.3 | NP_037179.1 | 2 | 956 | ||
XM_006241611.1 | XP_006241673.1 | 2 | 987 | |||
GenBank mRNA | M90308.1 | - | 1 | 706 | ||
S69160.1 | - | 1 | 617 | |||
S69161.1 | - | 1 | 617 | |||
X64630.1 | - | 2 | 884 | |||
X66726.1 | - | 2 | 957 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
