siRNA Details
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013056.1 | NP_037188.1 | 4 | 311 | ||
GenBank mRNA | X15143.1 | - | 2 | 192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013056.1 | NP_037188.1 | 4 | 311 | ||
GenBank mRNA | X15143.1 | - | 2 | 192 |