siRNA Details
| Gene Symbol: F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013057.2 | NP_037189.2 | 2 | 200 | ||
| GenBank mRNA | BC081846.1 | - | 2 | 200 | 7096602 | |
| U07619.1 | - | 2 | 167 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F3| Gene Symbol: F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013057.2 | NP_037189.2 | 2 | 200 | ||
| GenBank mRNA | BC081846.1 | - | 2 | 200 | 7096602 | |
| U07619.1 | - | 2 | 167 | |||