siRNA Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013059.1 | NP_037191.1 | 2 | 192 | ||
GenBank mRNA | J03572.1 | - | 2 | 192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013059.1 | NP_037191.1 | 2 | 192 | ||
GenBank mRNA | J03572.1 | - | 2 | 192 |