siRNA Details
| Gene Symbol: Agrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175754.1 | NP_786930.1 | 3 | 524 | ||
| XM_006239541.2 | XP_006239603.1 | 4 | 729 | |||
| XM_006239542.2 | XP_006239604.1 | 4 | 729 | |||
| XM_008764338.1 | XP_008762560.1 | 4 | 729 | |||
| XM_008764339.1 | XP_008762561.1 | 4 | 729 | |||
| XM_008764340.1 | XP_008762562.1 | 3 | 424 | |||
| GenBank mRNA | M64780.1 | - | 3 | 524 | ||

Agrn
Genomic Map